Anti-NEK6 Mouse Monoclonal Antibody [clone: OTI5B9]

Toimittaja: OriGene
ORIGTA500529EA 581 EUR
ORIGTA500529
Anti-NEK6 Mouse Monoclonal Antibody [clone: OTI5B9]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: NEK6 (NIMA-related kinase 6)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR