Anti-SLC2A5 Mouse Monoclonal Antibody [clone: OTI2C4]

Toimittaja: OriGene
ORIGTA500546EA 603 EUR
ORIGTA500546
Anti-SLC2A5 Mouse Monoclonal Antibody [clone: OTI2C4]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-1000; Immunochemistry: 1:50; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: SLC2A5 (solute carrier family 2 (facilitated glucose/fructose transporter), member 5)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR