Anti-CRYAB Mouse Monoclonal Antibody [clone: OTI2A1]

Toimittaja: OriGene
ORIGTA500550EA 603 EUR
ORIGTA500550
Anti-CRYAB Mouse Monoclonal Antibody [clone: OTI2A1]
Vasta-aineet
  • Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: CRYAB (crystallin, alpha B)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR