Anti-NEK6 Mouse Monoclonal Antibody [clone: OTI2H7]

Toimittaja: OriGene
ORIGTA500551EA 586 EUR
ORIGTA500551
Anti-NEK6 Mouse Monoclonal Antibody [clone: OTI2H7]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: NEK6 (NIMA-related kinase 6)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR