Anti-KDM4C Mouse Monoclonal Antibody [clone: OTI5B9]

Toimittaja: OriGene
ORIGTA500567EA 608 EUR
ORIGTA500567
Anti-KDM4C Mouse Monoclonal Antibody [clone: OTI5B9]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: KDM4C (lysine (K)-specific demethylase 4C)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR