Anti-PPP5C Mouse Monoclonal Antibody [clone: OTI5G5]

Toimittaja: OriGene
ORIGTA500593EA 603 EUR
ORIGTA500593
Anti-PPP5C Mouse Monoclonal Antibody [clone: OTI5G5]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:50; Immunofluorescence: 1:50-100; Flow Cytometry: 1:100

Type: Primary
Antigen: PPP5C (protein phosphatase 5, catalytic subunit)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR