Anti-PRKAR1A Mouse Monoclonal Antibody [clone: OTI6C7]

Toimittaja: OriGene
ORIGTA500641EA 609 EUR
ORIGTA500641
Anti-PRKAR1A Mouse Monoclonal Antibody [clone: OTI6C7]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: PRKAR1A (protein kinase, cAMP-dependent, regulatory, type I, alpha)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR