Anti-FCGR2A Mouse Monoclonal Antibody [clone: OTI19C10]

Toimittaja: OriGene
ORIGTA500647EA 603 EUR
ORIGTA500647
Anti-FCGR2A Mouse Monoclonal Antibody [clone: OTI19C10]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:200-1:1000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: FCGR2A (Fc fragment of IgG, low affinity IIa, receptor (CD32))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR