Anti-HSD17B10 Mouse Monoclonal Antibody [clone: OTI10B4]

Toimittaja: OriGene
ORIGTA500686EA 603 EUR
ORIGTA500686
Anti-HSD17B10 Mouse Monoclonal Antibody [clone: OTI10B4]
Vasta-aineet
  • Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50; Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: HSD17B10 (hydroxysteroid (17-beta) dehydrogenase 10)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR