Anti-HSPA1A Mouse Monoclonal Antibody [clone: OTI2C12]

Toimittaja: OriGene
ORIGTA500731EA 603 EUR
ORIGTA500731
Anti-HSPA1A Mouse Monoclonal Antibody [clone: OTI2C12]
Vasta-aineet
  • Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: HSPA1A (heat shock 70kDa protein 1A)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR