Anti-GBE1 Mouse Monoclonal Antibody [clone: OTI3C8]

Toimittaja: OriGene
ORIGTA500803EA 603 EUR
ORIGTA500803
Anti-GBE1 Mouse Monoclonal Antibody [clone: OTI3C8]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunoprecipitation: 2?g/500?l

Type: Primary
Antigen: GBE1 (glucan (1,4-alpha-), branching enzyme 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR