Anti-HNRNPF Mouse Monoclonal Antibody [clone: OTI5F5]

Toimittaja: OriGene
ORIGTA500804EA 608 EUR
ORIGTA500804
Anti-HNRNPF Mouse Monoclonal Antibody [clone: OTI5F5]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:50-100; Flow Cytometry: 1:100

Type: Primary
Antigen: HNRNPF (heterogeneous nuclear ribonucleoprotein F)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR