Anti-AKT2 Mouse Monoclonal Antibody [clone: OTI8G8]

Toimittaja: OriGene
ORIGTA500808EA 603 EUR
ORIGTA500808
Anti-AKT2 Mouse Monoclonal Antibody [clone: OTI8G8]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: AKT2 (v-akt murine thymoma viral oncogene homolog 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR