Anti-TTLL12 Mouse Monoclonal Antibody [clone: OTI6E4]

Toimittaja: OriGene
ORIGTA500816EA 581 EUR
ORIGTA500816
Anti-TTLL12 Mouse Monoclonal Antibody [clone: OTI6E4]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: TTLL12 (tubulin tyrosine ligase-like family, member 12)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR