Anti-PRL Mouse Monoclonal Antibody [clone: OTI2E4]

Toimittaja: OriGene
ORIGTA500817EA 581 EUR
ORIGTA500817
Anti-PRL Mouse Monoclonal Antibody [clone: OTI2E4]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: PRL (prolactin)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR