Anti-DCXR Mouse Monoclonal Antibody [clone: OTI4D11]

Toimittaja: OriGene
ORIGTA500820EA 603 EUR
ORIGTA500820
Anti-DCXR Mouse Monoclonal Antibody [clone: OTI4D11]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:50; Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: DCXR (dicarbonyl/L-xylulose reductase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR