Anti-HSPA1A Mouse Monoclonal Antibody [clone: OTI1H5]

Toimittaja: OriGene
ORIGTA500828EA 603 EUR
ORIGTA500828
Anti-HSPA1A Mouse Monoclonal Antibody [clone: OTI1H5]
Vasta-aineet
  • Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50

Type: Primary
Antigen: HSPA1A (heat shock 70kDa protein 1A)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR