Anti-PKMYT1 Mouse Monoclonal Antibody [clone: OTI2B4]
Toimittaja: OriGene
ORIGTA500839EA 603 EUR
ORIGTA500839
Anti-PKMYT1 Mouse Monoclonal Antibody [clone: OTI2B4]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: PKMYT1 (protein kinase, membrane associated tyrosine/threonine 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human