Anti-LOX Mouse Monoclonal Antibody [clone: OTI7E6]

Toimittaja: OriGene
ORIGTA500867EA 581 EUR
ORIGTA500867
Anti-LOX Mouse Monoclonal Antibody [clone: OTI7E6]
Vasta-aineet
Recommended Dilutions: Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: LOX (lysyl oxidase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR