Anti-ACO2 Mouse Monoclonal Antibody [clone: OTI7G4]

Toimittaja: OriGene
ORIGTA500873EA 608 EUR
ORIGTA500873
Anti-ACO2 Mouse Monoclonal Antibody [clone: OTI7G4]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100 Flow Cytometry: 1:100

Type: Primary
Antigen: ACO2 (aconitase 2, mitochondrial)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR