Anti-XRCC1 Mouse Monoclonal Antibody [clone: OTI2G8]

Toimittaja: OriGene
ORIGTA500879EA 581 EUR
ORIGTA500879
Anti-XRCC1 Mouse Monoclonal Antibody [clone: OTI2G8]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: XRCC1 (X-ray repair complementing defective repair in Chinese hamster cells 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR