Anti-IDH3A Mouse Monoclonal Antibody [clone: OTI2E9]
Toimittaja: OriGene
ORIGTA500912EA 603 EUR
ORIGTA500912
Anti-IDH3A Mouse Monoclonal Antibody [clone: OTI2E9]
Vasta-aineet
Recommended Dilutions: Immunofluorescence: 1:100 Flow Cytometry: 1:100
Type: Primary
Antigen: IDH3A (isocitrate dehydrogenase 3 (NAD+) alpha)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human