Anti-MTRF1L Mouse Monoclonal Antibody [clone: OTI1C8]

Toimittaja: OriGene
ORIGTA501012EA 603 EUR
ORIGTA501012
Anti-MTRF1L Mouse Monoclonal Antibody [clone: OTI1C8]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: MTRF1L (mitochondrial translational release factor 1-like)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR