Anti-TLDC1 Mouse Monoclonal Antibody [clone: OTI12B1]

Toimittaja: OriGene
ORIGTA501037EA 586 EUR
ORIGTA501037
Anti-TLDC1 Mouse Monoclonal Antibody [clone: OTI12B1]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunofluorescence: 1:100

Type: Primary
Antigen: TLDC1 (TBC/LysM-associated domain containing 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR