Anti-DNTTIP1 Mouse Monoclonal Antibody [clone: OTI4F7]

Toimittaja: OriGene
ORIGTA501130EA 603 EUR
ORIGTA501130
Anti-DNTTIP1 Mouse Monoclonal Antibody [clone: OTI4F7]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: DNTTIP1 (deoxynucleotidyltransferase, terminal, interacting protein 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR