Anti-GPI Mouse Monoclonal Antibody [clone: OTI7G10]

Toimittaja: OriGene
ORIGTA501136EA 603 EUR
ORIGTA501136
Anti-GPI Mouse Monoclonal Antibody [clone: OTI7G10]
Vasta-aineet
  • Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50

Type: Primary
Antigen: GPI (glucose-6-phosphate isomerase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR