Anti-ERCC1 Mouse Monoclonal Antibody [clone: OTI1C6]

Toimittaja: OriGene
ORIGTA501169EA 603 EUR
ORIGTA501169
Anti-ERCC1 Mouse Monoclonal Antibody [clone: OTI1C6]
Vasta-aineet
  • Recommended Dilutions: Western Blot: 1:1000; Immunofluorescence: 1:100; Flow Cytometry: 1:100; Immunochemistry: 1:50-1:150

Type: Primary
Antigen: ERCC1 (excision repair cross-complementation group 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR