Anti-SNX9 Mouse Monoclonal Antibody [clone: OTI1E4]

Toimittaja: OriGene
ORIGTA501252EA 603 EUR
ORIGTA501252
Anti-SNX9 Mouse Monoclonal Antibody [clone: OTI1E4]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: SNX9 (sorting nexin 9)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR