Anti-RANGAP1 Mouse Monoclonal Antibody [clone: OTI1B4]

Toimittaja: OriGene
ORIGTA501265EA 608 EUR
ORIGTA501265
Anti-RANGAP1 Mouse Monoclonal Antibody [clone: OTI1B4]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:1000-2000; Immunochemistry: 1:50; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: RANGAP1 (Ran GTPase activating protein 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR