Anti-MPP3 Mouse Monoclonal Antibody [clone: OTI8B11]

Toimittaja: OriGene
ORIGTA501306EA 581 EUR
ORIGTA501306
Anti-MPP3 Mouse Monoclonal Antibody [clone: OTI8B11]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Immunochemistry: 1:150

Type: Primary
Antigen: MPP3 (membrane protein, palmitoylated 3 (MAGUK p55 subfamily member 3))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR