Anti-MOB1B Mouse Monoclonal Antibody [clone: OTI1F6]

Toimittaja: OriGene
ORIGTA501362EA 616 EUR
ORIGTA501362
Anti-MOB1B Mouse Monoclonal Antibody [clone: OTI1F6]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100

Type: Primary
Antigen: MOB1B (MOB kinase activator 1B)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR