Anti-COX6B2 Mouse Monoclonal Antibody [clone: OTI1F5]
Toimittaja: OriGene
ORIGTA501377EA 608 EUR
ORIGTA501377
Anti-COX6B2 Mouse Monoclonal Antibody [clone: OTI1F5]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100
Type: Primary
Antigen: COX6B2 (cytochrome c oxidase subunit VIb polypeptide 2 (testis))
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human