Anti-MIOX Mouse Monoclonal Antibody [clone: OTI2F4]
Toimittaja: OriGene
ORIGTA501423EA 581 EUR
ORIGTA501423
Anti-MIOX Mouse Monoclonal Antibody [clone: OTI2F4]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Flow Cytometry: 1:100
Type: Primary
Antigen: MIOX (myo-inositol oxygenase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human