Anti-EFNA2 Mouse Monoclonal Antibody [clone: OTI3E3]

Toimittaja: OriGene
ORIGTA501461EA 609 EUR
ORIGTA501461
Anti-EFNA2 Mouse Monoclonal Antibody [clone: OTI3E3]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: EFNA2 (ephrin-A2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR