Anti-MOB3B Mouse Monoclonal Antibody [clone: OTI1G1]

Toimittaja: OriGene
ORIGTA501499EA 603 EUR
ORIGTA501499
Anti-MOB3B Mouse Monoclonal Antibody [clone: OTI1G1]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: MOB3B (MOB kinase activator 3B)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR