Anti-BTN1A1 Mouse Monoclonal Antibody [clone: OTI1G9]

Toimittaja: OriGene
ORIGTA501534EA 603 EUR
ORIGTA501534
Anti-BTN1A1 Mouse Monoclonal Antibody [clone: OTI1G9]
Vasta-aineet
  • Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: BTN1A1 (butyrophilin, subfamily 1, member A1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR