Anti-TRIM2 Mouse Monoclonal Antibody [clone: OTI2B10]

Toimittaja: OriGene
ORIGTA501606EA 608 EUR
ORIGTA501606
Anti-TRIM2 Mouse Monoclonal Antibody [clone: OTI2B10]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:1000-2000; Flow Cytometry: 1:100

Type: Primary
Antigen: TRIM2 (tripartite motif containing 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR