Anti-SULT2A1 Mouse Monoclonal Antibody [clone: OTI1D2]

Toimittaja: OriGene
ORIGTA501713EA 603 EUR
ORIGTA501713
Anti-SULT2A1 Mouse Monoclonal Antibody [clone: OTI1D2]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:1000-2000; Flow Cytometry: 1:100

Type: Primary
Antigen: SULT2A1 (sulfotransferase family, cytosolic, 2A, dehydroepiandrosterone (DHEA)-preferring, member 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR