Anti-PIK3AP1 Mouse Monoclonal Antibody [clone: OTI5A1]

Toimittaja: OriGene
ORIGTA501722EA 593 EUR
ORIGTA501722
Anti-PIK3AP1 Mouse Monoclonal Antibody [clone: OTI5A1]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: PIK3AP1 (phosphoinositide-3-kinase adaptor protein 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR