Anti-ARHGAP25 Mouse Monoclonal Antibody [clone: OTI11B9]
Toimittaja: OriGene
ORIGTA501750EA 603 EUR
ORIGTA501750
Anti-ARHGAP25 Mouse Monoclonal Antibody [clone: OTI11B9]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:150; Immunofluorescence: 1:100; Flow Cytometry: 1:100
Type: Primary
Antigen: ARHGAP25 (Rho GTPase activating protein 25)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human