Anti-AFP Mouse Monoclonal Antibody [clone: OTI1E8]
Toimittaja: OriGene
ORIGTA501783EA 603 EUR
ORIGTA501783
Anti-AFP Mouse Monoclonal Antibody [clone: OTI1E8]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500; Immunochemistry: 1:150; Flow Cytometry: 1:100
Type: Primary
Antigen: AFP (alpha-fetoprotein)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human