Anti-TCP11L2 Mouse Monoclonal Antibody [clone: OTI3H7]

Toimittaja: OriGene
ORIGTA501804EA 581 EUR
ORIGTA501804
Anti-TCP11L2 Mouse Monoclonal Antibody [clone: OTI3H7]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: TCP11L2 (t-complex 11, testis-specific-like 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR