Anti-GSTT2 Mouse Monoclonal Antibody [clone: OTI8A10]

Toimittaja: OriGene
ORIGTA501831EA 581 EUR
ORIGTA501831
Anti-GSTT2 Mouse Monoclonal Antibody [clone: OTI8A10]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:1000-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: GSTT2 (glutathione S-transferase theta 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR