Anti-ALDH1L1 Mouse Monoclonal Antibody [clone: OTI7B7]

Toimittaja: OriGene
ORIGTA501855EA 603 EUR
ORIGTA501855
Anti-ALDH1L1 Mouse Monoclonal Antibody [clone: OTI7B7]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: ALDH1L1 (aldehyde dehydrogenase 1 family, member L1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR