Anti-GSS Mouse Monoclonal Antibody [clone: OTI1H3]

Toimittaja: OriGene
ORIGTA501859EA 603 EUR
ORIGTA501859
Anti-GSS Mouse Monoclonal Antibody [clone: OTI1H3]
Vasta-aineet
  • Recommended Dilutions: Western Blot: 1:2000

Type: Primary
Antigen: GSS (glutathione synthetase)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR