Anti-EPM2AIP1 Mouse Monoclonal Antibody [clone: OTI1F3]

Toimittaja: OriGene
ORIGTA501931EA 603 EUR
ORIGTA501931
Anti-EPM2AIP1 Mouse Monoclonal Antibody [clone: OTI1F3]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:1000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: EPM2AIP1 (EPM2A (laforin) interacting protein 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR