Anti-RABL2A Mouse Monoclonal Antibody [clone: OTI4A8]

Toimittaja: OriGene
ORIGTA502018EA 608 EUR
ORIGTA502018
Anti-RABL2A Mouse Monoclonal Antibody [clone: OTI4A8]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: RABL2A (RAB, member of RAS oncogene family-like 2A)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR