Anti-ANXA3 Mouse Monoclonal Antibody [clone: OTI1A9]

Toimittaja: OriGene
ORIGTA502048EA 586 EUR
ORIGTA502048
Anti-ANXA3 Mouse Monoclonal Antibody [clone: OTI1A9]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:500-2000; Flow Cytometry: 1:100

Type: Primary
Antigen: ANXA3 (annexin A3)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR