Anti-CALCOCO2 Mouse Monoclonal Antibody [clone: OTI3F2]

Toimittaja: OriGene
ORIGTA502054EA 581 EUR
ORIGTA502054
Anti-CALCOCO2 Mouse Monoclonal Antibody [clone: OTI3F2]
Vasta-aineet
  • Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100

Type: Primary
Antigen: CALCOCO2 (calcium binding and coiled-coil domain 2)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR