Anti-SNTA1 Mouse Monoclonal Antibody [clone: OTI2B4]

Toimittaja: OriGene
ORIGTA502057EA 603 EUR
ORIGTA502057
Anti-SNTA1 Mouse Monoclonal Antibody [clone: OTI2B4]
Vasta-aineet
Recommended Dilutions: Western Blot: 1:2000; Immunofluorescence: 1:100; Flow Cytometry: 1:100

Type: Primary
Antigen: SNTA1 (syntrophin, alpha 1)
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR